APOH0430C0D7 Apollo Scientific Ltd

(2R)-2-Aminopropanamide hydrochloride

  • MDL number MFCD00039093
  • Cas Number : 71810-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-172
Synonym (2R)-2-Aminopropionamide hydrochloride, D-Alaninamide hydrochloride
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2r-2-aminopropanamide-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2r-2-aminopropanamide-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart