APOH0430C0D8 Apollo Scientific Ltd

Tropine

  • MDL number MFCD00005551
  • Cas Number : 120-29-6
  • Molecular Weight : 141.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-65
Synonym endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol, endo-3-Hydroxy-8-methyl-8-azabicyclo[3.2.1]octane
Density 1.04
Storage temp. Store at 2-8
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 232-234
Product Link https://store.apolloscientific.co.uk/product/tropine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/tropine
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart