APOH0430C0DA Apollo Scientific Ltd

4-(3-Chlorophenyl)but-3-en-2-one

  • MDL number MFCD00052851
  • Cas Number : 20766-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3-Chlorobenzylidene)acetone
Flash Point >110
Storage temp. Ambient temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 123-125/1mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=20216

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=20216
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart