APOH0430C0E1 Apollo Scientific Ltd

2-Methoxychalcone

  • MDL number MFCD00048035
  • Cas Number : 5416-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-57
Synonym 3-(2-Methoxyphenyl)-1-phenylprop-2-en-1-one
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23977

Safety Information

signalword Warning
pcodes P280;P304+P340;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23977
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart