APOH0430C0E3 Apollo Scientific Ltd

2'-Hydroxy-3,4,4'-trimethoxychalcone

  • MDL number MFCD00205556
  • Cas Number : 10493-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(3,4-Dimethoxyphenyl)-1-(2-hydroxy-4-methoxyphenyl)prop-2-en-1-one, 2-[3-(3,4-Dimethoxyphenyl)acryloyl]-5-methoxyphenol
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=23991

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=23991
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart