APOH0430C0E8 Apollo Scientific Ltd

2-Acetamido-2-deoxy-alpha-D-glucopyranose

  • MDL number MFCD00064359
  • Cas Number : 10036-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211
Synonym alpha-N-Acetyl-D-glucosamine, N-[(2S,3R,4R,5S,6R)-6-(Hydroxymethyl)-2,4,5-trihydroxytetrahydro-2H-pyran-3-yl]acetamide, 2-(Acetylamino)-2-deoxy-alpha-D-glucopyranose
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=25139

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=25139
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart