APOH0430C0EA Apollo Scientific Ltd

(3S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid, N-BOC protected

  • MDL number MFCD00143845
  • Cas Number : 78879-20-6
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3S)-2-(tert-butoxycarbonyl)-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid
Inchi Code 1S/C15H19NO4/c1-15(2,3)20-14(19)16-9-11-7-5-4-6-10(11)8-12(16)13(17)18/h4-7,12H,8-9H2,1-3H3,(H,17,18)/t12-/m0/s1
Inchi Key HFPVZPNLMJDJFB-LBPRGKRZSA-N

Description

Synonym Boc-L-Tic-OH
Purity 98%
Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3s-1234-tetrahydroisoquinoline-3-carboxylic-acid-n-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3s-1234-tetrahydroisoquinoline-3-carboxylic-acid-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart