APOH0430C0EB Apollo Scientific Ltd

3-Nitro-4-(pyridin-2-ylthio)cinammic acid

  • MDL number MFCD00173720
  • Cas Number : 175278-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-178
Synonym 3-{3-Nitro-4-[(pyridin-2-yl)thio]phenyl}acrylic acid, 3-{3-Nitro-4-[(pyridin-2-yl)thio]phenyl}prop-2-enoic acid
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26500

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26500
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart