APOH0430C0EC Apollo Scientific Ltd

Isopropyl (2E,4E,7S)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate

  • MDL number MFCD00867612
  • Cas Number : 65733-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Prop-2-yl (2E,4E,7S)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate, S-(+)-Methoprene
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26719

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26719
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart