APOH0430C0F0 Apollo Scientific Ltd

(E)-3-(2,3-Dihydrobenzo[b]furan-5-yl)acrylic acid

  • MDL number MFCD03002828
  • Cas Number : 203505-84-4
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-173
Synonym (2E)-3-(2,3-Dihydrobenzo[b]furan-5-yl)prop-2-enoic acid, (2E)-3-(2,3-Dihydro-1-benzofuran-5-yl)prop-2-enoic acid
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/e-3-23-dihydrobenzobfuran-5-ylacrylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/e-3-23-dihydrobenzobfuran-5-ylacrylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart