APOH0430C0F5 Apollo Scientific Ltd

1-Benzyl 2-methyl (2S,4S)-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride

  • MDL number MFCD06804628
  • Cas Number : 1212395-57-7
  • Molecular Weight : 314.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzyl 2-methyl (2S,4S)-4-amino-1,2-pyrrolidinedicarboxylate hydrochloride
Inchi Code 1S/C14H18N2O4.ClH/c1-19-13(17)12-7-11(15)8-16(12)14(18)20-9-10-5-3-2-4-6-10;/h2-6,11-12H,7-9,15H2,1H3;1H/t11-,12-;/m0./s1
Inchi Key VLLRSJJKBDFSMT-FXMYHANSSA-N

Description

Synonym (2S,4S)-4-Amino-1-[(benzyloxy)carbonyl]-2-(methoxycarbonyl)pyrrolidine hydrochloride, cis-4-Amino-L-proline methyl ester, N-CBZ protected hydrochloride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-benzyl-2-methyl-2s4s-4-aminopyrrolidine-12-dicarboxylate-hydrochloride

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-benzyl-2-methyl-2s4s-4-aminopyrrolidine-12-dicarboxylate-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart