APOH0430C11A Apollo Scientific Ltd

Methyl 2-[(dimethylamino)methylene]acetoacetate

  • MDL number MFCD08689663
  • Cas Number : 203186-56-5
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-67
Synonym Methyl 2-acetyl-3-(dimethylamino)acrylate, Methyl 2-[(dimethylamino)methylidene]-3-oxobutanoate
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-dimethylaminomethyleneacetoacetate

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/methyl-2-dimethylaminomethyleneacetoacetate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart