APOH0430C11C Apollo Scientific Ltd

Phenazine-1-carboxylic acid

  • MDL number MFCD00185184
  • Cas Number : 2538-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239-245
Synonym 1-Carboxyphenazine, 1-Carboxydibenzo[b,e]pyrazine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/phenazine-1-carboxylic-acid

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/phenazine-1-carboxylic-acid
pictograms GHS07;GHS08
hstatements H317;H319;H334
Availability details for
Quantity
  • Availability
  • Add to Cart