APOH0430C11D Apollo Scientific Ltd

(2S)-1-Benzylpyrrolidine-2-carboxylic acid

  • MDL number MFCD00235894
  • Cas Number : 31795-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Benzyl-L-proline
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38832

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38832
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart