APOH0430C11F Apollo Scientific Ltd

3-[(Dimethylamino)methylene]piperidin-4-one, N1-BOC protected

  • MDL number MFCD08443986
  • Cas Number : 157327-41-8
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 85-87
Iupac Name tert-butyl (3Z)-3-[(dimethylamino)methylene]-4-oxo-1-piperidinecarboxylate
Inchi Code 1S/C13H22N2O3/c1-13(2,3)18-12(17)15-7-6-11(16)10(9-15)8-14(4)5/h8H,6-7,9H2,1-5H3/b10-8-
Inchi Key YUSMZDVTEOAHDL-NTMALXAHSA-N

Description

Synonym tert-Butyl 3-[(dimethylamino)methylidene]-4-oxopiperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-[(dimethylamino)methylene]-4-oxopiperidine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 85-87
Product Link https://store.apolloscientific.co.uk/product/3-dimethylaminomethylenepiperidin-4-one-n1-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-dimethylaminomethylenepiperidin-4-one-n1-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart