APOH0430C126 Apollo Scientific Ltd

(2S)-2-Amino-3-(naphth-1-yl)propanoic acid

  • MDL number MFCD00066086
  • Cas Number : 55516-54-6
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 254
Synonym 3-(Naphth-1-yl)-L-alanine, (2S)-2-Amino-3-(naphth-1-yl)propionic acid, 1-[(2S)-2-Amino-2-carboxyethyl]naphthalene
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2s-2-amino-3-naphth-1-ylpropanoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2s-2-amino-3-naphth-1-ylpropanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart