APOH0430C127 Apollo Scientific Ltd

(2R)-2-Amino-3-(naphth-1-yl)propanoic acid

  • MDL number MFCD00038544
  • Cas Number : 78306-92-0
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2R)-2-amino-3-(1-naphthyl)propanoic acid
Inchi Code 1S/C13H13NO2/c14-12(13(15)16)8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7,12H,8,14H2,(H,15,16)/t12-/m1/s1
Inchi Key OFYAYGJCPXRNBL-GFCCVEGCSA-N

Description

Synonym (2R)-2-Amino-3-(naphth-1-yl)propionic acid, 3-(Naphth-1-yl)-D-alanine, 1-[(2R)-2-Amino-2-carboxyethyl]naphthalene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2r-2-amino-3-naphth-1-ylpropanoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2r-2-amino-3-naphth-1-ylpropanoic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart