APOH0430C134 Apollo Scientific Ltd

(6S)-1,4-Diazabicyclo[4.3.0]nonane

  • MDL number MFCD03787926
  • Cas Number : 93643-24-4
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 180
Iupac Name (8aS)-octahydropyrrolo[1,2-a]pyrazine
Inchi Code 1S/C7H14N2/c1-2-7-6-8-3-5-9(7)4-1/h7-8H,1-6H2/t7-/m0/s1
Inchi Key FTTATHOUSOIFOQ-ZETCQYMHSA-N

Description

Synonym (8aS)-Octahydropyrrolo[1,2-a]pyrazine
Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 180
Product Link https://store.apolloscientific.co.uk/product/6s-14-diazabicyclo430nonane

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6s-14-diazabicyclo430nonane
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart