APOH0430C135 Apollo Scientific Ltd

(6R)-1,4-Diazabicyclo[4.3.0]nonane

  • MDL number MFCD09752820
  • Cas Number : 96193-27-0
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (8aR)-Octahydropyrrolo[1,2-a]pyrazine
Physical Form Liquid-Oil
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 65-69/3mm
Product Link https://store.apolloscientific.co.uk/product/6r-14-diazabicyclo430nonane

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6r-14-diazabicyclo430nonane
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart