APOH0430C138 Apollo Scientific Ltd

(2S)-2-Isobutylpiperazine, N1-BOC protected

  • MDL number MFCD03787916
  • Cas Number : 674792-06-4
  • Molecular Weight : 242.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (2S)-2-isobutyl-1-piperazinecarboxylate
Inchi Code 1S/C13H26N2O2/c1-10(2)8-11-9-14-6-7-15(11)12(16)17-13(3,4)5/h10-11,14H,6-9H2,1-5H3/t11-/m0/s1
Inchi Key WXGGVOBNOVOVAM-NSHDSACASA-N

Description

Synonym tert-Butyl (2S)-2-(2-methylprop-1-yl)piperazine-1-carboxylate, (2S)-1-(tert-Butoxycarbonyl)-2-isobutylpiperazine
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2s-2-isobutylpiperazine-n1-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2s-2-isobutylpiperazine-n1-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart