APOH0430C141 Apollo Scientific Ltd

Methyl (2R,4S)-4-hydroxypyrrolidine-2-carboxylate hydrochloride

  • MDL number MFCD00135747
  • Cas Number : 481704-21-6
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl (2R,4S)-4-hydroxy-2-pyrrolidinecarboxylate hydrochloride
Inchi Code 1S/C6H11NO3.ClH/c1-10-6(9)5-2-4(8)3-7-5;/h4-5,7-8H,2-3H2,1H3;1H/t4-,5+;/m0./s1
Inchi Key KLGSHNXEUZOKHH-UYXJWNHNSA-N

Description

Synonym trans-4-Hydroxy-D-proline methyl ester hydrochloride, (2R,4S)-4-Hydroxy-2-(methoxycarbonyl)pyrrolidine hydrochloride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2r4s-4-hydroxypyrrolidine-2-carboxylate-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2r4s-4-hydroxypyrrolidine-2-carboxylate-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart