APOH0430C14A Apollo Scientific Ltd

Ethyl (2E)-pent-2-enoate

  • MDL number MFCD06204561
  • Cas Number : 24410-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl trans-pent-2-enoate
Density 0.9086
Storage temp. Ambient temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45832

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45832
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart