APOH0430C14F Apollo Scientific Ltd

2-Acetamido-2-deoxy-D-mannopyranose

  • MDL number MFCD00046758
  • Cas Number : 3615-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Acetyl-D-mannosamine, 2-(Acetylamino)-2-deoxy-D-mannopyranose
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47968

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47968
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart