APOH0430C152 Apollo Scientific Ltd

O-Methyl-L-threonine, N-BOC protected

  • MDL number MFCD00076990
  • Cas Number : 48068-25-3
  • Molecular Weight : 233.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2S,3R)-2-[(tert-Butoxycarbonyl)amino]-3-methoxybutanoic acid
Purity 98
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/o-methyl-l-threonine-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/o-methyl-l-threonine-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart