APOH0430C155 Apollo Scientific Ltd

(2S)-3-(Acetylthio)-2-methylpropanoic acid

  • MDL number MFCD00038563
  • Cas Number : 76497-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 5
Synonym (2S)-3-(Acetylsulphanyl)-2-methylpropanoic acid, (2S)-3-(Acetylthio)-2-methylpropionic acid
Density 1.178
Flash Point >112.8
Storage temp. Ambient temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 131-132/2.25mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=50625

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=50625
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart