APOH0430C157 Apollo Scientific Ltd

Ethyl 2-benzoyl-3-(dimethylamino)acrylate

  • MDL number MFCD00120477
  • Cas Number : 66129-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-65
Synonym Ethyl 2-benzoyl-3-(dimethylamino)prop-2-enoate, Ethyl 2-[(dimethylamino)methylene]-3-oxo-3-phenylpropanoate
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 150-155/0.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51927

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51927
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart