APOH0430C158 Apollo Scientific Ltd

Methyl pent-2-enoate

  • MDL number MFCD00137611
  • Cas Number : 818-59-7
  • Molecular Weight : 114.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl (2E)-2-pentenoate
Inchi Code 1S/C6H10O2/c1-3-4-5-6(7)8-2/h4-5H,3H2,1-2H3/b5-4+
Inchi Key MBAHGFJTIVZLFB-SNAWJCMRSA-N

Description

Synonym Methyl 3-ethylacrylate
Purity 97%
Flash Point 81-82/45mm
Storage temp. Store at -20
Physical Form Liquid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-pent-2-enoate

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-pent-2-enoate
pictograms GHS02;GHS07
hstatements H225;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart