APOH0430C15C Apollo Scientific Ltd

1-(3,5-Dichlorophenyl)ethylamine hydrochloride

  • MDL number MFCD18705305
  • Cas Number : 321318-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63906

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=63906
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart