APOH0430C15D Apollo Scientific Ltd

(4R)-2,2-Dioxido-4-methyl-1,2,3-oxathiazolidine, N-BOC protected

  • MDL number MFCD09864994
  • Cas Number : 454248-53-4
  • Molecular Weight : 237.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-124
Synonym tert-Butyl (4R)-4-methyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide, (4R)-3-(tert-Butoxycarbonyl)-4-methyl-1,2,3-oxathiazolidine 2,2-dioxide
Purity 95
Storage temp. Store at 2-8
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4r-22-dioxido-4-methyl-123-oxathiazolidine-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4r-22-dioxido-4-methyl-123-oxathiazolidine-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart