APOH0430C160 Apollo Scientific Ltd

5-Bromo-2-iodonitrobenzene

  • MDL number MFCD11505492
  • Cas Number : 112671-42-8
  • Molecular Weight : 327.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-89
Synonym 4-Bromo-1-iodo-2-nitrobenzene
Purity 99
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-iodonitrobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-iodonitrobenzene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart