APOH0430C169 Apollo Scientific Ltd

Trimethylsilyl N-(trimethylsilyl)trifluoroacetimidate

  • MDL number MFCD00008269
  • Cas Number : 25561-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -10
Synonym BSTFA, Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate, Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, N,O-Bis(trimethylsilyl)trifluoroacetamide
Purity 99
Density 0.969
Flash Point 24
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 45-50/14mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=404

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=404
pictograms GHS02;GHS05
hstatements H226;H314
Availability details for
Quantity
  • Availability
  • Add to Cart