APOH0430C16B Apollo Scientific Ltd

2-Chloro-6-fluorocinnamic acid

  • MDL number MFCD00051582
  • Cas Number : 392-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-172
Synonym 3-(2-Chloro-6-fluorophenyl)acrylic acid
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=838

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=838
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart