APOH0430C175 Apollo Scientific Ltd

2-Fluoro-3-(trifluoromethyl)cinnamic acid

  • MDL number MFCD00236291
  • Cas Number : 237069-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-[2-Fluoro-3-(trifluoromethyl)phenyl]prop-2-enoic acid, 3-[2-Fluoro-3-(trifluoromethyl)phenyl]acrylic acid, 3-(2-Carboxyethenyl)-2-fluorobenzotrifluoride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-trifluoromethylcinnamic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-trifluoromethylcinnamic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart