APOH0430C180 Apollo Scientific Ltd

3-(Perfluorohexyl)prop-2-en-1-ol

  • MDL number MFCD00042355
  • Cas Number : 38550-47-9
  • Molecular Weight : 376.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1H,1H,2H,3H-Perfluoronon-2-en-1-ol
Purity 98
Density 1.65
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 87-89/10mm
Product Link https://store.apolloscientific.co.uk/product/3-perfluorohexylprop-2-en-1-ol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-perfluorohexylprop-2-en-1-ol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart