APOH0430C187 Apollo Scientific Ltd

1-[3-(Trifluoromethyl)phenyl]but-1-en-3-one

  • MDL number MFCD09998085
  • Cas Number : 80992-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 115/2.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5390

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5390
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart