APOH0430C18E Apollo Scientific Ltd

(1S)-(-)-1-[3,5-Bis(trifluoromethyl)phenyl]ethan-1-ol

  • MDL number MFCD03093011
  • Cas Number : 225920-05-8
  • Molecular Weight : 258.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (S)-(-)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl alcohol
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 83/11mm
Product Link https://store.apolloscientific.co.uk/product/1s-1-35-bistrifluoromethylphenylethan-1-ol

Safety Information

signalword Warning
pcodes P271;P280;P261
MSDS https://store.apolloscientific.co.uk/product/1s-1-35-bistrifluoromethylphenylethan-1-ol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart