APOH0430C18F Apollo Scientific Ltd

(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine

  • MDL number MFCD03093012
  • Cas Number : 127733-47-5
  • Molecular Weight : 257.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/r-1-35-bistrifluoromethylphenylethylamine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/r-1-35-bistrifluoromethylphenylethylamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart