APOH0430C190 Apollo Scientific Ltd

(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine

  • MDL number MFCD03093013
  • Cas Number : 127733-40-8
  • Molecular Weight : 257.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.1
Flash Point 66
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 63.5-64.5
Product Link https://store.apolloscientific.co.uk/product/s-1-35-bistrifluoromethylphenylethylamine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/s-1-35-bistrifluoromethylphenylethylamine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart