APOH0430C194 Apollo Scientific Ltd

(1S)-1-[4-(Trifluoromethyl)phenyl]ethylamine

  • MDL number MFCD03093009
  • Cas Number : 84499-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (S)-alpha-Methyl-4-(trifluoromethyl)benzylamine
Purity 98
Density 1.03
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 76/20mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6241

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6241
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart