APOH0430C195 Apollo Scientific Ltd

(1R)-1-(2-Fluorophenyl)ethan-1-ol

  • MDL number MFCD03092991
  • Cas Number : 162427-79-4
  • Molecular Weight : 140.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (R)-2-Fluoro-alpha-methylbenzyl alcohol
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1r-1-2-fluorophenylethan-1-ol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1r-1-2-fluorophenylethan-1-ol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart