APOH0430C197 Apollo Scientific Ltd

(1R)-1-(2-Fluorophenyl)ethylamine

  • MDL number MFCD03092993
  • Cas Number : 185545-90-8
  • Molecular Weight : 139.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.00
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1r-1-2-fluorophenylethylamine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1r-1-2-fluorophenylethylamine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart