APOH0430C199 Apollo Scientific Ltd

2,3,4,5,6-Pentafluorochalcone

  • MDL number MFCD00483321
  • Cas Number : 15269-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Pentafluorophenyl)-1-phenylprop-2-en-1-one, 3-(Perfluorophenyl)-1-phenylprop-2-en-1-one
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6360

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6360
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart