APOH0430C1A3 Apollo Scientific Ltd

(1R)-1-(Trifluoromethyl)ethylamine hydrochloride

  • MDL number MFCD07779095
  • Cas Number : 177469-12-4
  • Molecular Weight : 149.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2R)-1,1,1-Trifluoropropan-2-amine hydrochloride, (2R)-2-Amino-1,1,1-trifluoropropane hydrochloride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1r-1-trifluoromethylethylamine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1r-1-trifluoromethylethylamine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart