APOH0430C1A4 Apollo Scientific Ltd

(1R)-1-(3-Fluorophenyl)ethylamine

  • MDL number MFCD06761859
  • Cas Number : 761390-58-3
  • Molecular Weight : 139.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 72
Iupac Name (1R)-1-(3-fluorophenyl)ethanamine
Inchi Code 1S/C8H10FN/c1-6(10)7-3-2-4-8(9)5-7/h2-6H,10H2,1H3/t6-/m1/s1
Inchi Key ASNVMKIDRJZXQZ-ZCFIWIBFSA-N

Description

Melting Point 72
Synonym (R)-3-Fluoro-alpha-methylbenzylamine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1r-1-3-fluorophenylethylamine

Safety Information

signalword Danger
pcodes P280;P260;P271
MSDS https://store.apolloscientific.co.uk/product/1r-1-3-fluorophenylethylamine
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart