APOH0430C1A8 Apollo Scientific Ltd

4-(Trifluoromethoxy)-2-(trifluoromethyl)aniline hydrochloride

  • MDL number MFCD19105214
  • Cas Number : 1588441-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63905

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=63905
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart