APOH0680A4A9 Apollo Scientific Ltd

2,5-Dihydro-1-oxy-2,2,5,5-tetramethylpyrrole-3-carboxylic acid, free radical 250mg pack

  • MDL number MFCD00041847
  • Cas Number : 2154-67-8
  • Molecular Weight : 184.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Oxy-2,2,5,5-tetramethyl-3-pyrroline-3-carboxylic acid, free radical
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 202
Product Link https://store.apolloscientific.co.uk/product/25-dihydro-1-oxy-2255-tetramethylpyrrole-3-carboxylic-acid-free-radical

Safety Information

signalword Warning
pcodes P271;P261;P280
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart