APOH0680A670 Apollo Scientific Ltd

Dimethyl cyclohexane-1,2-dicarboxylate

  • MDL number MFCD00043768
  • Cas Number : 1687-29-2
  • Molecular Weight : 200.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name dimethyl (1R,2S)-1,2-cyclohexanedicarboxylate
density 1.12
Inchi Code 1S/C10H16O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h7-8H,3-6H2,1-2H3/t7-,8+
Inchi Key AIACXWOETVLBIA-OCAPTIKFSA-N

Description

Purity 96%
Density 1.12
Flash Point 111
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/dimethyl-cyclohexane-12-dicarboxylate

Safety Information

signalword Warning
pcodes P280;P302+P352
MSDS https://store.apolloscientific.co.uk/product/dimethyl-cyclohexane-12-dicarboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart