APOH0680BB04 Apollo Scientific Ltd

3-Methyl-1H-pyrazole-5-carbohydrazide

  • MDL number MFCD00068101
  • Cas Number : 40535-14-6
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 158-160
Iupac Name 3-methyl-1H-pyrazole-5-carbohydrazide
Inchi Code 1S/C5H8N4O/c1-3-2-4(9-8-3)5(10)7-6/h2H,6H2,1H3,(H,7,10)(H,8,9)
Inchi Key FWNHUZOBVQZERU-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 158-160
Product Link https://store.apolloscientific.co.uk/product/3-methyl-1h-pyrazole-5-carbohydrazide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-methyl-1h-pyrazole-5-carbohydrazide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart