APOH0680BDA3 Apollo Scientific Ltd

4-Propylbenzene-1,3-diol

  • MDL number MFCD00020120
  • Cas Number : 18979-60-7
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 82.5
Iupac Name 4-propyl-1,3-benzenediol
Inchi Code 1S/C9H12O2/c1-2-3-7-4-5-8(10)6-9(7)11/h4-6,10-11H,2-3H2,1H3
Inchi Key DJDHQJFHXLBJNF-UHFFFAOYSA-N

Description

Purity 97%
Flash Point 286.9
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 82.5
Product Link https://store.apolloscientific.co.uk/product/4-propylbenzene-13-diol

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/4-propylbenzene-13-diol
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart