APOH0680BDA7 Apollo Scientific Ltd

(Pyridin-4-yl)acetone

  • MDL number MFCD00129043
  • Cas Number : 6304-16-1
  • Molecular Weight : 135.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/pyridin-4-ylacetone

Safety Information

signalword Warning
pcodes P280;P302+P352;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/pyridin-4-ylacetone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart